Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027640.5 | NP_001022811.1 | 1 - 2 | 208 | 150 | |
NM_001381690.1 | NP_001367960.1 | 5 - 6 | 566 | 150 | ||
NM_001384004.1 | 20 - 21 | 2967 | 150 | |||
NM_001384005.1 | 2 - 3 | 240 | 150 | |||
NM_001384006.1 | 1 - 2 | 210 | 150 | |||
NM_064789.5 | NP_497190.1 | 6 - 7 | 999 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |