Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027459.6 | NP_001022630.2 | 1 - 2 | 122 | 59 | |
NM_064931.6 | NP_497332.4 | 1 - 2 | 120 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |