Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pes-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064950.6 | NP_497351.3 | 7 - 8 | 1020 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |