Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp-60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129251.4 | NP_001122723.1 | 4 - 5 | 997 | 72 | |
NM_001306670.3 | NP_001293599.1 | 2 - 3 | 628 | 72 | ||
NM_065028.8 | NP_497429.1 | 5 - 6 | 1563 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |