Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129227.4 | NP_001122699.2 | 10 - 11 | 2236 | 100 | |
NM_001267920.3 | NP_001254849.1 | 10 - 11 | 2236 | 100 | ||
NM_065077.6 | NP_497478.2 | 10 - 11 | 2245 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |