Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065116.1 | NP_497517.1 | 2 - 3 | 222 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |