Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065250.3 | NP_497651.2 | 7 - 8 | 850 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |