Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ral-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267937.3 | NP_001254866.1 | 2 - 3 | 460 | 55 | |
NM_001267938.2 | NP_001254867.1 | 1 - 2 | 52 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |