Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267939.3 | NP_001254868.1 | 23 - 24 | 4002 | 113 | |
NM_001267940.3 | NP_001254869.1 | 1 - 2 | 386 | 113 | ||
NM_065293.6 | NP_497694.3 | 24 - 25 | 4233 | 113 | ||
NM_065294.6 | NP_497695.3 | 22 - 23 | 3706 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |