Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065331.4 | NP_497732.1 | 1 - 2 | 190 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |