Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065332.4 | NP_497733.1 | 1 - 2 | 199 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |