Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026024.3 | NP_001021195.1 | 1 - 2 | 105 | 83 | |
NM_001384056.1 | 2 - 3 | 183 | 83 | |||
GenBank mRNA | AF241846.1 | - | 2 - 3 | 205 | 83 | |
AF241847.1 | - | 1 - 2 | 127 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |