Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026047.5 | NP_001021218.1 | 15 - 16 | 2035 | 92 | |
NM_001381807.1 | NP_001366876.1 | 15 - 16 | 2023 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |