Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065358.5 | NP_497759.1 | 2 - 3 | 182 | 71 | |
GenBank mRNA | AF134806.1 | - | 2 - 3 | 184 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |