Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065408.8 | NP_497809.1 | 6 - 7 | 1058 | 84 | |
GenBank mRNA | X75564.1 | - | 1069 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |