Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wht-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322779.3 | NP_001309589.1 | 7 | 1288 | 155 | |
NM_001322780.3 | NP_001309591.1 | 5 - 6 | 1072 | 155 | ||
NM_001322781.3 | NP_001309590.1 | 5 - 6 | 1114 | 155 | ||
NM_065424.3 | NP_497825.2 | 7 - 8 | 1377 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |