Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267961.2 | NP_001254890.1 | 3 - 4 | 452 | 107 | |
NM_001267962.2 | NP_001254891.1 | 2 - 3 | 315 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |