Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377920.3 | NP_001364559.1 | 31 - 32 | 7413 | 78 | |
NM_065538.6 | NP_497939.2 | 30 - 31 | 7278 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |