Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065539.6 | NP_497940.2 | 4 - 5 | 1001 | 91 | |
GenBank mRNA | U19861.2 | - | 4 - 5 | 996 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |