Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pal-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026038.6 | NP_001021209.1 | 5 - 6 | 737 | 59 | |
NM_001026039.6 | NP_001021210.1 | 5 - 6 | 781 | 59 | ||
NM_001267992.4 | NP_001254921.1 | 3 - 4 | 457 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |