Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065638.6 | NP_498039.1 | 1 - 2 | 64 | 85 | |
NM_065639.6 | NP_498040.1 | 1 - 2 | 137 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |