Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqe-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382869.1 | 4 - 5 | 298 | 63 | ||
NM_065734.8 | NP_498135.2 | 4 - 5 | 304 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |