Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065761.5 | NP_498162.2 | 1 - 2 | 519 | 83 | |
GenBank mRNA | AB110419.1 | - | 1 - 2 | 508 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |