Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025981.3 | NP_001021152.1 | 4 - 5 | 752 | 73 | |
NM_001025982.5 | NP_001021153.1 | 7 - 8 | 889 | 73 | ||
GenBank mRNA | AB016838.4 | - | 4 - 5 | 752 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |