Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgg-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065827.2 | NP_498228.1 | 2 - 3 | 275 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |