Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312927.3 | NP_001299856.1 | 4 - 5 | 1758 | 109 | |
NM_065835.8 | NP_498236.1 | 6 - 7 | 1953 | 109 | ||
GenBank mRNA | AF013951.1 | - | 6 - 7 | 1920 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |