Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065899.7 | NP_498300.2 | 3 - 4 | 365 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Caenorhabditis elegans Probe spans exons |