Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027821.6 | NP_001022992.1 | 12 - 13 | 3424 | 94 | |
NM_065910.7 | NP_498311.2 | 12 - 13 | 3409 | 94 | ||
GenBank mRNA | AF469173.1 | - | 12 - 13 | 3362 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |