Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wht-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066024.3 | NP_498425.2 | 9 - 10 | 1551 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |