Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kbp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306710.2 | NP_001293639.1 | 1 - 2 | 86 | 140 |
Gene Symbol: K04C2.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027489.5 | NP_001022660.2 | 1 - 2 | 85 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |