Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027384.6 | NP_001022555.1 | 2 - 3 | 375 | 90 | |
GenBank mRNA | U01303.1 | - | 2 - 3 | 375 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |