Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276780.3 | NP_001263709.1 | 2 - 3 | 181 | 112 | |
NM_001276782.3 | NP_001263711.1 | 2 - 3 | 191 | 112 | ||
NM_001276783.3 | NP_001263712.1 | 2 - 3 | 181 | 112 | ||
NM_001276785.3 | NP_001263714.1 | 2 - 3 | 181 | 112 | ||
NM_001276786.3 | NP_001263715.1 | 2 - 3 | 181 | 112 | ||
NM_001276787.4 | NP_001263716.1 | 2 - 3 | 181 | 112 | ||
NM_001276788.3 | NP_001263717.1 | 2 - 3 | 181 | 112 | ||
NM_001276789.3 | NP_001263718.1 | 2 - 3 | 181 | 112 | ||
NM_066269.6 | NP_498670.2 | 2 - 3 | 181 | 112 | ||
GenBank mRNA | JQ990141.1 | - | 2 - 3 | 181 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |