Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066294.7 | NP_498695.1 | 4 - 5 | 553 | 107 | |
GenBank mRNA | AF277990.1 | - | 4 - 5 | 568 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |