Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047424.3 | NP_001040889.1 | 9 - 10 | 1459 | 101 | |
NM_001379835.1 | NP_001367402.1 | 9 - 10 | 1471 | 101 | ||
GenBank mRNA | AJ276027.1 | - | 9 - 10 | 1505 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |