Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066484.2 | NP_498885.1 | 4 - 5 | 1132 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |