Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066486.8 | NP_498887.2 | 9 - 10 | 2008 | 68 | |
GenBank mRNA | U34661.1 | - | 9 - 10 | 2087 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |