Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066533.6 | NP_498934.1 | 3 - 4 | 594 | 76 | |
GenBank mRNA | AB110823.1 | - | 2 - 3 | 657 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |