Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027844.4 | NP_001023015.1 | 3 - 4 | 335 | 101 | |
GenBank mRNA | AF269694.1 | - | 3 - 4 | 329 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |