Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066693.6 | NP_499094.1 | 4 - 5 | 684 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |