Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mog-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066811.9 | NP_499212.1 | 5 - 6 | 2913 | 109 | |
GenBank mRNA | AF120269.1 | - | 5 - 6 | 2928 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |