Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066854.6 | NP_499255.1 | 2 - 3 | 351 | 93 | |
GenBank mRNA | AF031935.1 | - | 2 - 3 | 351 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |