Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379897.1 | NP_001367241.1 | 7 - 8 | 1880 | 68 | |
NM_001379898.1 | NP_001366696.1 | 7 - 8 | 1641 | 68 | ||
NM_001379899.1 | NP_001367186.1 | 6 - 7 | 1556 | 68 | ||
NM_001379900.2 | NP_001367113.1 | 7 - 8 | 1529 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |