Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rle-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379916.1 | NP_001366782.1 | 4 - 5 | 1915 | 71 | |
NM_001379917.1 | NP_001367296.1 | 4 - 5 | 1917 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |