Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsa-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027715.3 | NP_001022886.1 | 2 - 3 | 247 | 62 | |
NM_001027716.4 | NP_001022887.1 | 2 - 3 | 176 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |