Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pha-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027719.5 | NP_001022890.1 | 2 - 3 | 440 | 140 | |
NM_001027720.2 | NP_001022891.1 | 3 - 4 | 350 | 140 | ||
GenBank mRNA | X73845.1 | - | 471 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |