Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wah-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276800.3 | NP_001263729.1 | 3 - 4 | 530 | 70 | |
NM_067163.5 | NP_499564.2 | 5 - 6 | 1217 | 70 | ||
NR_101859.1 | 4 - 5 | 1504 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |