Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027660.3 | NP_001022831.1 | 3 - 4 | 581 | 63 | |
NM_067288.5 | NP_499689.1 | 3 - 4 | 297 | 63 | ||
GenBank mRNA | AF109378.1 | - | 3 - 4 | 259 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |