Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379972.2 | NP_001367103.1 | 2 - 3 | 602 | 105 | |
NM_001379973.1 | NP_001367104.1 | 2 - 3 | 599 | 105 | ||
NM_001383018.1 | 3 - 4 | 660 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |