Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067323.3 | NP_499724.1 | 1 - 2 | 98 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |