Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lis-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067354.7 | NP_499755.1 | 6 - 7 | 723 | 66 | |
GenBank mRNA | AF164430.1 | - | 6 - 7 | 714 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |